Learn More
Medchemexpress LLC 3-sulfo-taurocholic acid disodium salt | 71781-33-4 | 99.0% | 639.73 | C26H43NNa2O10S2 | 1mg
SDP
Catalog No. 5000270343
Encompass_Preferred
3-Sulfo-taurocholic Acid Disodium Salt (3-Sulfocholyl Taurine TCA3S) is a metabolite of the conjugated bile acid taurocholic acid Plasma levels of 3-Sulfo-taurocholic Acid Disodium Salt are elevated in wild-type and Sortilin 1 (Sort1) knockout mice at 6 hours following bile duct ligation (BDL) and are further elevated in Sort1 knockout mice at 24 hours post-BDL
Catalog No. 50-002-70343 Supplier Medchemexpress LLC Supplier No. HYW7470721MG
3-Sulfo-taurocholic Acid Disodium Salt (3-Sulfocholyl Taurine TCA3S) is a metabolite of the conjugated bile acid taurocholic acid Plasma levels of 3-Sulfo-taurocholic Acid Disodium Salt are elevated in wild-type and Sortilin 1 (Sort1) knockout mice at 6 hours following bile duct ligation (BDL) and are further elevated in Sort1 knockout mice at 24 hours post-BDL