Learn More
Paper Thermometer Co 3-TEMP THERMLBL 150F 160F 170F
Supplier:  Paper Thermometer Co TL3160
Encompass
3-Temp THERMOLABEL® 150°F 160°F 170°F 16 LABELSWaterproof 3-Temp Thermolabel® self-adhering dishmachine thermometers are available in packages of 16 labels each. Our waterproof 3-Temp Thermolabel® is a popular irreversible registering temperature indicator for dishwasher sanitation (and HACCP compliance). These temperature strips for dishwashers can be used in...
Catalog No. NC3244433