Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(tert-butoxycarbonyl)-3-azabicyclo[4.1.0]heptane-1-carboxylic acid | 1239421-67-0 | 95.0% | 241.29 g/mol | C12H19NO4 | 100 MG
SDP

Catalog No. 5000210489
Encompass_Preferred

3-(tert-butoxycarbonyl)-3-azabicyclo[4.1.0]heptane-1-carboxylic acid is a bicyclic carboxylic acid used as a drug intermediate in the synthesis of various active compounds.

  • Bicyclic carboxylic acid scaffold suitable for synthetic derivatization.
  • Molecular weight 241.29 g/mol.
  • CAS number 1239421-67-0 for unambiguous identification.
  • Reported high purity for research-grade material.
  • Available in small-scale packages for laboratory use.

Catalog No. 50-002-10489 Supplier Medchemexpress LLC Supplier No. HYW068934100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(tert-butoxycarbonyl)-3-azabicyclo[4.1.0]heptane-1-carboxylic acid is a bicyclic carboxylic acid used as a drug intermediate in the synthesis of various active compounds.

  • Bicyclic carboxylic acid scaffold suitable for synthetic derivatization.
  • Molecular weight 241.29 g/mol.
  • CAS number 1239421-67-0 for unambiguous identification.
  • Reported high purity for research-grade material.
  • Available in small-scale packages for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.