Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[(tert-Butoxycarbonyl)amino]phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B42795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4279-5G 5 g
B42791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4279-5G Supplier TCI America Supplier No. B42795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 380430-68-2
Melting Point 168°C
Molecular Formula C11H16BNO4
Quantity 5 g
MDL Number MFCD03411945
Synonym 3-boc-aminophenyl boronic acid, 3-n-boc-amino phenylboronic acid, 3-tert-butoxycarbonylamino phenylboronic acid, n-boc-3-aminophenylboronic acid, 3-tert-butoxycarbonyl amino phenylboronic acid
InChI Key CWLNHPXWZRALFS-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)NC(=O)OC(C)(C)C)(O)O
IUPAC Name [3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
Molecular Weight (g/mol) 237.062
PubChem CID 2773228
Formula Weight 237.06
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.