Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. B47265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4726-5G 5 g
B47261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4726-5G Supplier TCI America Supplier No. B47265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 220210-56-0
Molecular Formula C11H15BO4
Molecular Weight (g/mol) 222.05
MDL Number MFCD01630855
InChI Key HVJDVHCPCSZDSR-UHFFFAOYSA-N
PubChem CID 2773299
IUPAC Name {3-[(tert-butoxy)carbonyl]phenyl}boronic acid
SMILES CC(C)(C)OC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Synonym 3-tert-butoxycarbonylphenylboronic acid, 3-tert-butoxycarbonyl phenylboronic acid, 3-t-butoxycarbonyl phenylboronic acid, tert-butyl 3-boronobenzoate, 3-tert-butoxycarbonyl phenyl boronic acid
Formula Weight 222.05
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.