Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-tert-Butyl-2-hydroxybenzaldehyde

Catalog No. 4818745G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-187-45G 5 g
48-187-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-187-45G Supplier Sigma Aldrich Supplier No. 4818745G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-tert-butyl-2-hydroxybenzaldehyde is a mono-tert-butyl substituted 2-hydroxybenzaldehyde that can be synthesized using ethylbromide and 2-tert-butylphenol.3-tert-butyl-2-hydroxybenzaldehyde may be used in the synthesis of:
  • 5-bromo-3-tert-butyl-2-hydroxybenzaldehyde
  • 3-tert-butyl-5-(chloro-methyl)-2- hydroxybenzaldehyde
  • Chiral schiff base-titanium alkoxide catalysts
  • 2 -(3-tert-butyl -2-hydroxybenzyl)aminomethylpyridine

Specifications

CAS 24623-65-2
Density 1.041 g/mL (at 25°C (literature))
Boiling Point 78°C to 79°C (1 mmHg)
Molecular Formula C11H14O2
Refractive Index n20/D 1.544 (literature)
Linear Formula (CH3)3 CC6H3(OH)CHO
Quantity 5 g
MDL Number MFCD00778877
Synonym 3-tert-Butylsalicylaldehyde
Molecular Weight (g/mol) 178.23
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.