Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-tert-butyl-4-methoxyphenol | 88-32-4 | MFCD00057667 | 99.7% | 180.24 g/mol | C11H16O2 | 10 G
SDP

Catalog No. 5000301243
Encompass_Preferred

3-tert-Butyl-4-methoxyphenol (CAS 88-32-4) is an aromatic small molecule provided as a high-purity research reagent commonly used as a PROTAC linker and building block in small-molecule studies. It is supplied as a solid suitable for synthetic chemistry and assay development.

  • High purity: 99.69% (reported).
  • Molecular formula: C11H16O2.
  • Molecular weight: 180.24 g/mol.
  • Available in multiple pack sizes, including 10 g.
  • Suitable for linker synthesis and general research applications.

Catalog No. 50-003-01243 Supplier Medchemexpress LLC Supplier No. HYW02436510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-tert-Butyl-4-methoxyphenol (CAS 88-32-4) is an aromatic small molecule provided as a high-purity research reagent commonly used as a PROTAC linker and building block in small-molecule studies. It is supplied as a solid suitable for synthetic chemistry and assay development.

  • High purity: 99.69% (reported).
  • Molecular formula: C11H16O2.
  • Molecular weight: 180.24 g/mol.
  • Available in multiple pack sizes, including 10 g.
  • Suitable for linker synthesis and general research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.