Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-tert-Butyl-5-ethyltoluene 98.0+%, TCI America™
SDP

Catalog No. B145910ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B145910ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B145910ML Supplier TCI America Supplier No. B145910ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6630-01-9
Molecular Formula C13H20
Molecular Weight (g/mol) 176.30
MDL Number MFCD00026954
InChI Key URDKTSKLGNWKDB-UHFFFAOYSA-N
PubChem CID 23114
IUPAC Name 1-tert-butyl-3-ethyl-5-methylbenzene
SMILES CCC1=CC(=CC(=C1)C)C(C)(C)C

Specifications

Color Yellow
Boiling Point 71°C
Quantity 10 mL
Formula Weight 176.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-tert-Butyl-5-ethyltoluene
EINECSNumber (3)-3427
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.