Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(tert-Butyldimethylsiloxy)glutaric anhydride, 95%

Catalog No. AAH6122706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6122706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6122706 Supplier Thermo Scientific Chemicals Supplier No. H6122706

missing translation for 'validMainframeMsg'

CAS: 91424-40-7 | C11H20O4Si | 244.362 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91424-40-7
Molecular Formula C11H20O4Si
Molecular Weight (g/mol) 244.362
MDL Number MFCD00075235
InChI Key RXAJGRHLLRGVSB-UHFFFAOYSA-N
Synonym 3-tert-butyldimethylsilyloxy glutaric anhydride, 4-tert-butyldimethylsilyl oxy oxane-2,6-dione, 3-tert-butyldimethylsilyl oxy pentanedioic anhydride, 3-tert-butyldimethylsilyloxy glutaric anhydride,, 4-tert-butyldimethylsilyl oxy dihydro-2h-pyran-2,6 3h-dione, acmc-209rbg, 4-tert-butyl dimethyl silyl oxyoxane-2,6-dione, 3-tert-butyldimethylsilyloxy glutaricanhydride, 3-tert-butyldimethylsiloxy pentanedioic anhydride, 3-tert-butyldimethyl silyl oxy glutaric anhydride
PubChem CID 4141040
IUPAC Name 4-[tert-butyl(dimethyl)silyl]oxyoxane-2,6-dione
SMILES CC(C)(C)[Si](C)(C)OC1CC(=O)OC(=O)C1

Specifications

Melting Point 79°C to 81°C
Density 1.030 g/mL
Flash Point >113°C (235°F)
Refractive Index 1.451
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 244.36
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-(tert-Butyldimethylsiloxy)glutaric anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.