Learn More
Hogentogler & Co Inc 3" Test Sieve, No. 14 Mesh, Half Height, Stainless Frame - Stainless Cloth
SDP
Catalog No. NC1545381
Encompass_Preferred
Model 1708 is a 3" test sieve with a No. 14 mesh. This sieve is constructed with a Stainless Frame - Stainless Cloth and is Half Height.
Catalog No. NC1545381 Supplier Hogentogler & Co Inc Supplier No. 1708
Model 1708 is a 3" test sieve with a No. 14 mesh. This sieve is constructed with a Stainless Frame - Stainless Cloth and is Half Height.