Learn More
Medchemexpress LLC 3-Thiazolidinepropanoic acid, 5-(5-chloro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)-4-oxo-2-thioxo- | 1426138-42-2 | 98.0% | 368.82 g/mol | C14H9ClN2O4S2 | 200 MG
SDP
Catalog No. 5000247373
Encompass_Preferred
FX1 is a small-molecule inhibitor of the transcriptional repressor BCL6, used as a research tool to probe BCL6-mediated transcriptional regulation and apoptosis. It is supplied as a solid and is suitable for biochemical and cellular studies.
  • Inhibits BCL6 with IC50 ≈ 35 μM
  • High purity (98.0%) suitable for research applications
  • Solid form, pink to red color
  • Molecular formula C14H9ClN2O4S2; molecular weight 368.82 g/mol
  • Available in multiple pack sizes including 200 MG
  • Powder storage: -20°C (3 years) or 4°C (2 years)
  • Intended for biochemical assays and cellular pathway studies
Catalog No. 50-002-47373 Supplier Medchemexpress LLC Supplier No. HY102027200MG
FX1 is a small-molecule inhibitor of the transcriptional repressor BCL6, used as a research tool to probe BCL6-mediated transcriptional regulation and apoptosis. It is supplied as a solid and is suitable for biochemical and cellular studies.
  • Inhibits BCL6 with IC50 ≈ 35 μM
  • High purity (98.0%) suitable for research applications
  • Solid form, pink to red color
  • Molecular formula C14H9ClN2O4S2; molecular weight 368.82 g/mol
  • Available in multiple pack sizes including 200 MG
  • Powder storage: -20°C (3 years) or 4°C (2 years)
  • Intended for biochemical assays and cellular pathway studies