Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Thienylboronic Acid

Catalog No. 4368445G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-684-45G 5 g
43-684-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-684-45G Supplier Sigma Aldrich Supplier No. 4368445G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Thienylboronic Acid

Specifications

CAS 6165-69-1
Melting Point 164°C to 169°C
Molecular Formula C4H5BO2S
Linear Formula C4H5BO2S
MDL Number MFCD00151851
Quantity 5 g
Synonym (Thiophene-3-yl)boronic acid; 3-Thiopheneboric acid; 3-Thiopheneboronic acid; 3-Thiophenylboronic acid; Thiophene-3-boronic acid
Molecular Weight (g/mol) 127.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.