Learn More
Sigma Aldrich 3-Thiophen-2-yl-piperazine-1-carboxylic acid ethyl ester
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Diisooctylthiophosphinic acid is an application specific reagent.diisooctylthiophosphinic acid is suitable for use in the colorimetric determination of airborne lead
- It may be used as chelating agent for supercritical fluid extraction (sfe) of metals, for subsequent icp-ms analyses of mercury, cadmium and lead in sediment
Specifications
Specifications
| CAS | 85803-50-5 |
| Molecular Formula | C11H16N2O2S |
| Linear Formula | C11H16N2O2S |
| MDL Number | MFCD06245431 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 240.32 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.