Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T19755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1975-5G 5 g
T19751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1975-5G Supplier TCI America Supplier No. T19755G
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 3-Thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 6165-69-1
Color White-Yellow
Molecular Formula C4H5BO2S
Quantity 5 g
MDL Number MFCD00151851
Synonym 3-thiopheneboronic acid, thiophene-3-boronic acid, 3-thienylboronic acid, thiophene 3-boronic acid, thiophene-3-boronicacid
InChI Key QNMBSXGYAQZCTN-UHFFFAOYSA-N
SMILES B(C1=CSC=C1)(O)O
IUPAC Name thiophen-3-ylboronic acid
Molecular Weight (g/mol) 127.952
PubChem CID 581760
Formula Weight 127.95
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.