Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiophenecarbonitrile, 94%, Thermo Scientific™

Catalog No. AC263160010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC263160010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC263160010 Supplier Thermo Scientific Chemicals Supplier No. 263160010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1641-09-4
Molecular Formula C5H3NS
Molecular Weight (g/mol) 109.15
MDL Number MFCD00151852
InChI Key GSXCEVHRIVLFJV-UHFFFAOYSA-N
Synonym 3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h
PubChem CID 74231
IUPAC Name thiophene-3-carbonitrile
SMILES C1=CSC=C1C#N

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Colorless
Boiling Point 82°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Refractive Index 1.5595 to 1.5615
Quantity 1 g
Formula Weight 109.15
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 3-Thiophenecarbonitrile, 94%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-687-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.