Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Thiophenecarboxylic acid, 99%

Catalog No. p-199862
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC243110010 1 g Glass bottle
AC243110050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC243110010 Supplier Thermo Scientific Chemicals Supplier No. 243110010
Only null left
Add to Cart

CAS: 88-13-1 | C5H4O2S | 128.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-13-1
Molecular Formula C5H4O2S
Molecular Weight (g/mol) 128.15
MDL Number MFCD00005467
InChI Key YNVOMSDITJMNET-UHFFFAOYSA-N
Synonym 3-thiophenecarboxylic acid, 3-thenoic acid, 3-thiophenezoic acid, beta-thiophenic acid, 3-thienylcarboxylic acid, 3-thiophene carboxylic acid, 3-carboxythiophene, beta-thiophenecarboxylic acid, 3-thiophenoic acid, unii-6v3012q6be
PubChem CID 6918
IUPAC Name thiophene-3-carboxylic acid
SMILES C1=CSC=C1C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 137°C to 139°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 18, 292
Fieser 01,1012
Merck Index 15, 9430
Solubility Information Solubility in water: 4.3g/L (25°C)
Formula Weight 128.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Thiophenecarboxylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-802-5
RTECSNumber XM8330300
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.