Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Tolylboronic acid | 17933-03-8 | 135.96 | 100 G
SDP

Catalog No. 5000460279
Encompass_Preferred

3-Tolylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • For research use only
  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US (may vary elsewhere)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Documents available: data sheet, COA, SDS, handling instructions

Catalog No. 50-004-60279 Supplier Medchemexpress LLC Supplier No. HYW007381100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Tolylboronic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • For research use only
  • Appearance: solid
  • Color: white to off-white
  • Shipping: room temperature in continental US (may vary elsewhere)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Documents available: data sheet, COA, SDS, handling instructions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.