Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Triallylsilyl)propyl Methacrylate (stabilized with MEHQ) 92.0+%, TCI America™
SDP

Catalog No. T32295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32295G Supplier TCI America Supplier No. T32295G

Chemical Identifiers

CAS 1990509-31-3
Molecular Formula C16H26O2Si
Synonym Methacrylic Acid 3-(Triallylsilyl)propyl Ester

Specifications

Quantity 5 g
Formula Weight 278.47
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Triallylsilyl)propyl Methacrylate (stabilized with MEHQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.