Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Triethoxysilyl)propionitrile

Catalog No. 12537725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-537-725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-537-725ML Supplier Sigma Aldrich Supplier No. 12537725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Triethoxysilyl)propionitrile (TESPN) is an organosilyl sol gel that can be used as a silane based coupling agent to improve the adhesion between metal-polymeric composite. It is non-toxic and cost efficient which makes it a suitable alternative to the existing chromate treatment of metals. It can be used in surface modification of silica based surfaces.TESPN can functionalize silica nanoparticles for the fabrication of solid state electrolyte based dye sensitized solar cells (DSSCs) and for the development of PVDf based ultrafiltration membranes for antifouling applications. It can also be used in the synthesis of SBA-15 for the formation of mesoporous silica for drug delivery applications.

Specifications

CAS 919-31-3
Density 0.979 g/mL (at 25°C)
Boiling Point 224°C
Molecular Formula C9H19NO3Si
Refractive Index n20/D 1.414 (literature)
Linear Formula (C2H5O)3 SiCH2CH2CN
Quantity 25 mL
MDL Number MFCD00019851
Synonym (2-Cyanoethyl)triethoxysilane
Molecular Weight (g/mol) 217.34
Percent Purity 97%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.