Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Triethoxysilyl)Propyl Isocyanate

Catalog No. 41336425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-336-425ML 25 mL
41-336-4100ML 100 mL
41-336-4500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 41-336-425ML Supplier Sigma Aldrich Supplier No. 41336425ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(triethoxysilyl)propyl isocyanate (TEPI) was used as a spacer reagent to prepare cellulose based chiral stationary phases (CSPs). TEPI may be used to modify 2-hydroxynicotinic acid to prepare luminescent hybrid materials.2 Luminescent inorganic-organic hybrid materials were prepared by grafting the isocynate with the ortho aminobenzoic acid derivative. TEPI may also be used to prepare 5-(N,N-bis-3-(triethoxysilyl)propyl)ureyl-1,10- phenanthroline (phen-Si).

Specifications

CAS 24801-88-5
Density 0.999 g/mL at 25°C
Boiling Point 283°C
Molecular Formula C10H21NO4Si
Refractive Index n20/D 1.42
Linear Formula (C2H5O)3Si(CH2)3NCO
MDL Number MFCD00051459
Quantity 25 mL
Synonym Triethoxy(3-isocyanatopropyl)silane; (3-Isocyanatopropyl)triethoxysilane
Molecular Weight (g/mol) 247.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.