Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-3-(Trifluoroacetyl)Camphor

Catalog No. 1890061G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
800 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-900-61G 800 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 18-900-61G Supplier Sigma Aldrich Supplier No. 1890061G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+)-3-(Trifluoroacetyl)camphor reacts with suitable chiral ketone precursors to generate chiral dioxiranes in situ, which can catalyse the direct asymmetric epoxidation of unfunctionalized olefins.

Specifications

CAS 563-43-9
Density 0.927 g/mL at 25°C
Molecular Formula C2H5AlCl2
Refractive Index n20/D 1.451
Linear Formula CH3CH2AlCl2
MDL Number MFCD00000457
Quantity 800 g
Synonym 3-(Trifluoroacetyl)-D-camphor
Molecular Weight (g/mol) 126.95 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.