Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-3-(Trifluoroacetyl)Camphor

Catalog No. 2983441G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-834-41G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 29-834-41G Supplier Sigma Aldrich Supplier No. 2983441G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-3-(Trifluoroacetyl)Camphor

Specifications

CAS 64536-78-3
Melting Point 175°C
Molecular Formula C31H50F6IrNP2
Linear Formula C31H50F6IrNP2
Refractive Index n20/D 1.451
MDL Number MFCD00075097
Quantity 250 mg
Synonym 3-(Trifluoroacetyl)-L-camphor
Molecular Weight (g/mol) 804.89 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.