Learn More
Accela Chembio Inc 3-(trifluoromethoxy)iodobenzene | 25g | 198206-33-6 | MFCD01090992 | 97+% | D: 1.863 | Shelf Life: 1260 Days | Light Sensitive
Catalog No. 50252142
Encompass_Preferred
3-(trifluoromethoxy)iodobenzene | 25g | 198206-33-6 | MFCD01090992 | 97+% | D: 1.863 | Shelf Life: 1260 Days | Light Sensitive
Catalog No. 50-252-142 Supplier Accela Chembio Inc Supplier No. SY01616325G
3-(trifluoromethoxy)iodobenzene | 25g | 198206-33-6 | MFCD01090992 | 97+% | D: 1.863 | Shelf Life: 1260 Days | Light Sensitive