Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethoxy)phenylacetic Acid 95.0+%, TCI America™
SDP

Catalog No. T24065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24065G Supplier TCI America Supplier No. T24065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 203302-97-0
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD00082480
InChI Key NFZQVADYFXRRPM-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy phenylacetic acid, 3-trifluoromethoxyphenylacetic acid, 2-3-trifluoromethoxy phenyl acetic acid, benzeneacetic acid, 3-trifluoromethoxy, 3-trifluoromethoxy phenylaceticacid, 3-trifluoromethoxy phenyl acetic acid, 3-trifluoromethoxyphenylaceticacid, pubchem4221, acmc-209f8q
PubChem CID 2777316
IUPAC Name 2-[3-(trifluoromethoxy)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)OC(F)(F)F)CC(=O)O

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
Formula Weight 220.15
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethoxy)phenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.