Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. T26005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26005G 5 g
T26001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26005G Supplier TCI America Supplier No. T26005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22245-83-6
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.099
MDL Number MFCD00153194
InChI Key JHDCDEHVUADNKQ-UHFFFAOYSA-N
Synonym 3-(Trifluoromethyl)-2-pyridinone, 2-Hydroxy-3-(trifluoromethyl)pyridine
PubChem CID 3678411
IUPAC Name 3-(trifluoromethyl)-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1)C(F)(F)F

Specifications

Melting Point 155°C
Color White
Quantity 5 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.