Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Trifluoromethyl-3-azetidinol hydrochloride, 96%, Thermo Scientific™

Catalog No. AAH66227MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66227MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66227MD Supplier Thermo Scientific Chemicals Supplier No. H66227MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 848192-96-1
Molecular Formula C4H7ClF3NO
Molecular Weight (g/mol) 177.551
MDL Number MFCD13248564
InChI Key BWGPIPWVIWINLD-UHFFFAOYSA-N
Synonym 3-trifluoromethyl azetidin-3-ol hydrochloride, 3-trifluoromethyl azetidin-3-ol hcl, 3-trifluoromethyl-3-azetidinol hydrochloride, 3-azetidinol, 3-trifluoromethyl-, hydrochloride, 3-hydroxy-3-trifluoromethyl azetidine hydrochloride, c4h6f3no.hcl, 3-trifluoromethyl-azetidin-3-ol hydrochloride, 3-trifluoromethyl ;azetidin-3-ol hydrochloride, 3-trifluoromethyl azetidin-3-ol-hydrogen chloride 1/1
PubChem CID 46735152
IUPAC Name 3-(trifluoromethyl)azetidin-3-ol;hydrochloride
SMILES C1C(CN1)(C(F)(F)F)O.Cl

Specifications

Color White to Yellow
Quantity 250 mg
Formula Weight 177.55
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 3-Trifluoromethyl-3-azetidinol hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.