Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(Trifluoromethyl)benzaldehyde | 454-89-7 | MFCD00003373 | 99.3% | C8H5F3O | 1 ML
SDP

Catalog No. 5000433736
Encompass_Preferred

3-(Trifluoromethyl)benzaldehyde is a key intermediate in organic synthesis, used to synthesize 2,3-di- and 2,2,3-trisubstituted-3-methoxycarbonyl-γ-butyrolactones. Its derivatives have demonstrated antitumor effects against various cancers, including nasopharyngeal carcinoma, leukemia, and breast cancer.

  • High purity
  • Molecular weight: 174.12
  • Chemical formula: C8H5F3O
  • CAS number: 454-89-7
  • Appearance: Colorless to light yellow liquid
  • Key intermediate in organic synthesis
  • Derivatives show antitumor effects

Catalog No. 50-004-33736 Supplier Medchemexpress LLC Supplier No. HY7657410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(Trifluoromethyl)benzaldehyde is a key intermediate in organic synthesis, used to synthesize 2,3-di- and 2,2,3-trisubstituted-3-methoxycarbonyl-γ-butyrolactones. Its derivatives have demonstrated antitumor effects against various cancers, including nasopharyngeal carcinoma, leukemia, and breast cancer.

  • High purity
  • Molecular weight: 174.12
  • Chemical formula: C8H5F3O
  • CAS number: 454-89-7
  • Appearance: Colorless to light yellow liquid
  • Key intermediate in organic synthesis
  • Derivatives show antitumor effects

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.