Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. T29251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29251G 1 g
T29255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29251G Supplier TCI America Supplier No. T29251G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 672-58-2
Molecular Formula C7H6F3NO2S
Molecular Weight (g/mol) 225.185
MDL Number MFCD00042421
InChI Key ZUTVRDMZQSHCID-UHFFFAOYSA-N
Synonym 3-trifluoromethyl benzenesulfonamide, 3-trifluoromethyl benzene-1-sulfonamide, 3-trifluoromethyl benzenesulphonamide, m-trifluoromethylbenzenesulfonamide, benzenesulfonamide, 3-trifluoromethyl, pubchem11763, maybridge3_000947, 3-sulphamoylbenzotrifluoride, ksc493o8t, 3-trifluoromethyl-benzenesulfonamide
PubChem CID 244076
IUPAC Name 3-(trifluoromethyl)benzenesulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)C(F)(F)F

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 225.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Trifluoromethyl)benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.