Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)benzoic acid, Thermo Scientific™

Catalog No. DP01925EA
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
DP01925EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DP01925EA Supplier Thermo Scientific Chemicals Supplier No. DP01925EA
This item is not returnable. View return policy

Specifications

CAS 454-92-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H5F3O2
Linear Formula CF3C6H4CO2H
MDL Number MFCD00002519
Quantity 10g
Synonym 3-trifluoromethyl benzoic acid, m-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-m-toluic acid, 3-carboxybenzotrifluoride, 3-trifluoromethyl-benzoic acid, benzoic acid, 3-trifluoromethyl, m-trifluoromethyl benzoic acid, 3-trifluoromethyl benzoicacid, m-carboxybenzotrifluoride, a,a,a-trifluoro-m-toluic acid
InChI Key FQXQBFUUVCDIRK-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC(=C1)C(F)(F)F
IUPAC Name 3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 190.12
PubChem CID 9963
ChEBI CHEBI:60695
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethyl)benzoic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.