Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Trifluoromethyl)cyclohexanol, mixture of cis/trans isomers

Catalog No. MNO0000021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MNO0000021G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MNO0000021G Supplier Sigma Aldrich Supplier No. MNO0000021G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-isopropyl-n-methyl-tert-butylamine is a tertiary amine
  • Bh3.thf stabilized by n-isopropyl n-methyl tert-butylamine is useful for the carbonyl reductions.n-isopropyl-n-methyl-tert-butylamine may be used to stabilize the borane-thf solutions
  • It may also be used for the synthesis of rotaxane, via sonogashira coupling

Specifications

CAS 454-63-7
Molecular Formula C7H11F3O
Linear Formula C7H11F3O
MDL Number MFCD00102143
Quantity 5 mL
Molecular Weight (g/mol) 168.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.