Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)phenylacetone 98.0+%, TCI America™
SDP

Catalog No. T16935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16935G 5 g
T169325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16935G Supplier TCI America Supplier No. T16935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21906-39-8
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.176
MDL Number MFCD00000397
InChI Key JPHQCDCEBDRIOL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl propan-2-one, m-trifluoromethyl phenylacetone, 1-3-trifluoromethyl phenyl acetone, 3-trifluormethyl phenylacetone, 1-3-trifluoromethyl phenyl-2-propanone, m-trifluoromethylphenylacetone, 2-propanone, 1-3-trifluoromethyl phenyl
PubChem CID 89101
IUPAC Name 1-[3-(trifluoromethyl)phenyl]propan-2-one
SMILES CC(=O)CC1=CC(=CC=C1)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 202.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethyl)phenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.