Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193825 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17931G 1 g
T1793-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17931G Supplier TCI America Supplier No. T17931G
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 3-(Trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 1423-26-3
Color White-Yellow
Molecular Formula C7H6BF3O2
Quantity 1 g
MDL Number MFCD00151854
Synonym 3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
InChI Key WOAORAPRPVIATR-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC(=C1)C(F)(F)F
IUPAC Name [3-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 189.93
PubChem CID 2734388
Formula Weight 189.93
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.