Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-3-(Trifluoromethyl)piperidine, 98%, Thermo Scientific™

Catalog No. AC454820010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454820010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454820010 Supplier Thermo Scientific Chemicals Supplier No. 454820010
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 768-31-0
Molecular Formula C6H11F3N
Molecular Weight (g/mol) 154.16
MDL Number MFCD02183561
InChI Key JOHFJTBDUSVGQB-RXMQYKEDSA-O
Synonym 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl
PubChem CID 2760776
IUPAC Name 3-(trifluoromethyl)piperidine
SMILES FC(F)(F)[C@@H]1CCC[NH2+]C1

Specifications

Boiling Point 128.0°C to 130.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 1 g
Formula Weight 153.15
Percent Purity 98%
Chemical Name or Material (±)-3-(Trifluoromethyl)piperidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.