Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-(Trifluoromethyl)propiophenone 97.0+%, TCI America™
SDP

Catalog No. T24045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24045G 5 g
T240425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24045G Supplier TCI America Supplier No. T24045G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 1533-03-5
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.176
MDL Number MFCD00039227
InChI Key UBTPNKXHYILGJU-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl propiophenone, m-trifluoromethylpropiophenone, 1-3-trifluoromethyl phenyl propan-1-one, 1-propanone, 1-3-trifluoromethyl phenyl, propiophenone, 3'-trifluoromethyl, 3-trifluoromethyl propiophenone, 1-3-trifluoromethyl-phenyl-propan-1-one, 1-3-trifluoromethyl phenyl-1-propanone
PubChem CID 137064
IUPAC Name 1-[3-(trifluoromethyl)phenyl]propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 202.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-(Trifluoromethyl)propiophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.