Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trifluoromethyl)styrene, 97+%, stabilized, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
402-24-4
C9H7F3
172.15
MFCD00075481
ARHOUOIHKWELMD-UHFFFAOYSA-N
3-trifluoromethyl styrene,1-trifluoromethyl-3-vinylbenzene,1-ethenyl-3-trifluoromethyl benzene,3-cf3c6h4ch=ch2,3-trifluoromethylstyren,3-vinylbenzotrifluoride,m-trifluoromethylstyrene,pubchem15477,3-trifluoromethylstyrene,1-trifluoromethyl-3-vinyl-benzene
520986
1-ethenyl-3-(trifluoromethyl)benzene
C=CC1=CC(=CC=C1)C(F)(F)F
402-24-4
C9H7F3
172.15
MFCD00075481
ARHOUOIHKWELMD-UHFFFAOYSA-N
3-trifluoromethyl styrene,1-trifluoromethyl-3-vinylbenzene,1-ethenyl-3-trifluoromethyl benzene,3-cf3c6h4ch=ch2,3-trifluoromethylstyren,3-vinylbenzotrifluoride,m-trifluoromethylstyrene,pubchem15477,3-trifluoromethylstyrene,1-trifluoromethyl-3-vinyl-benzene
520986
1-ethenyl-3-(trifluoromethyl)benzene
C=CC1=CC(=CC=C1)C(F)(F)F
Boiling Point 64.5°C (40.0mmHg)
Flash Point 42°C
Specific Gravity 1.161
Density 1.1610g/mL
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Packaging Glass bottle
Color Colorless
Infrared Spectrum Authentic
Quantity 1g
Stabilizer about 100ppm 4-tert.-butylcatechol
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97% min. (GC)
Linear Formula CH2CHC6H4CF3
Formula Weight 172.15
Physical Form Liquid
Percent Purity 97+%
Chemical Name or Material 3-(Trifluoromethyl)styrene
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning