Learn More
Medchemexpress LLC 3- Trimethoxysilyl p 50g
SDP
Catalog No. 5000206794
Encompass_Preferred
3-(Trimethoxysilyl)propyl methacrylate is a reactive monomer and silane coupling agent 3-(Trimethoxysilyl)propyl methacrylate can be used to prepare nanoparticles[1][2]
Catalog No. 50-002-06794 Supplier Medchemexpress LLC Supplier No. HYW10542650G
3-(Trimethoxysilyl)propyl methacrylate is a reactive monomer and silane coupling agent 3-(Trimethoxysilyl)propyl methacrylate can be used to prepare nanoparticles[1][2]