Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Trimethoxysilyl)propyl acrylate

Catalog No. 4751495ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-514-95ML 5 mL
47-514-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-514-95ML Supplier Sigma Aldrich Supplier No. 4751495ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Trimethoxysilyl)propyl acrylate is a reactive monomer that is majorly used as a silane coupling agent for the formation of hybrid compounds. It facilitates the formation of a hydrophobic surface by lowering the surface tension and increasing the wettability.

Specifications

CAS 4369-14-6
Density 1.055 g/mL (at 25°C)
Boiling Point 68°C (0.4 mmHg) (literature)
Molecular Formula C9H18O5Si
Refractive Index n20/D 1.429 (literature)
Linear Formula H2C=CHCO2(CH2)3 Si(OCH3)3
Quantity 5 mL
MDL Number MFCD00054803
Synonym (3-Acryloyloxypropyl)trimethoxysilane
Molecular Weight (g/mol) 234.32
Percent Purity 92%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.