Learn More
Sigma Aldrich Fine Chemicals Biosciences 3-Trimethoxysilylpropyl me
Catalog No. 502897575
Encompass
3-(Trimethoxysilyl)propyl methacrylate (TMSPMA) is a hydrophobic monomer. The presence of an additional functional group of acrylate facilitates TMSPMA for grafting polymerization on plastic surfaces. 3-(Trimethoxysilyl)propyl methacrylate undergoes first-order kinetic polymerization to form polyTMSPMA which is a moisture stable polymer.
Catalog No. 50-289-7575 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M651425ML
3-(Trimethoxysilyl)propyl methacrylate (TMSPMA) is a hydrophobic monomer. The presence of an additional functional group of acrylate facilitates TMSPMA for grafting polymerization on plastic surfaces. 3-(Trimethoxysilyl)propyl methacrylate undergoes first-order kinetic polymerization to form polyTMSPMA which is a moisture stable polymer.