Learn More
Medchemexpress LLC 3-(trimethylsilyl)propanoic acid-d4 sodium | 24493-21-8 | MFCD00002762 | 98.0% | 172.27 | C6H9D4NaO2Si | 10 MG
SDP
Catalog No. 5000284703
Encompass_Preferred
3-(Trimethylsilyl)propanoic acid-d4 sodium is the deuterium-labeled sodium salt used as an internal standard and tracer in analytical chemistry. It provides a stable reference signal for NMR spectroscopy and is also suitable for quantitative analyses by GC-MS and LC-MS, with high isotopic enrichment and purity.
  • Deuterium-labeled sodium salt suitable as an internal standard for 1H NMR.
  • Water- and deuterium oxide-soluble reference for reliable chemical shift calibration.
  • High isotopic enrichment and 98.0% purity support accurate quantitative analysis.
  • Applicable as a tracer in GC-MS and LC-MS workflows.
  • Supplied in small, accurately weighed packages for analytical use.
Catalog No. 50-002-84703 Supplier Medchemexpress LLC Supplier No. HYW70263710MG
3-(Trimethylsilyl)propanoic acid-d4 sodium is the deuterium-labeled sodium salt used as an internal standard and tracer in analytical chemistry. It provides a stable reference signal for NMR spectroscopy and is also suitable for quantitative analyses by GC-MS and LC-MS, with high isotopic enrichment and purity.
  • Deuterium-labeled sodium salt suitable as an internal standard for 1H NMR.
  • Water- and deuterium oxide-soluble reference for reliable chemical shift calibration.
  • High isotopic enrichment and 98.0% purity support accurate quantitative analysis.
  • Applicable as a tracer in GC-MS and LC-MS workflows.
  • Supplied in small, accurately weighed packages for analytical use.