Learn More
Medchemexpress LLC 3-(Trimethylsilyl)propanoic acid-d4 (sodium) | 24493-21-8 | MFCD00002762 | 98.0% | 172.27 | C6H9D4NaO2Si | 25 MG
SDP
Catalog No. 5000284666
Encompass_Preferred
3-(Trimethylsilyl)propanoic acid-d4 sodium is a deuterium-labeled sodium salt used primarily as an internal standard for nuclear magnetic resonance (NMR) and as a tracer or internal standard in quantitative mass-spectrometry methods. The compound is typically supplied as a white crystalline powder with reported molecular weight 172.27, CAS number 24493-21-8, and high purity for analytical use.
  • Deuterium-labeled sodium salt suitable for NMR internal standard use.
  • Applicable as an internal standard or tracer for quantitative NMR, GC-MS, and LC-MS.
  • Molecular weight 172.27 and CAS 24493-21-8 for unambiguous identification.
  • High purity (98.0%) to support accurate quantitative analyses.
  • White crystalline powder; store sealed at 4 °C and protect from moisture.
Catalog No. 50-002-84666 Supplier Medchemexpress LLC Supplier No. HYW70263725MG
3-(Trimethylsilyl)propanoic acid-d4 sodium is a deuterium-labeled sodium salt used primarily as an internal standard for nuclear magnetic resonance (NMR) and as a tracer or internal standard in quantitative mass-spectrometry methods. The compound is typically supplied as a white crystalline powder with reported molecular weight 172.27, CAS number 24493-21-8, and high purity for analytical use.
  • Deuterium-labeled sodium salt suitable for NMR internal standard use.
  • Applicable as an internal standard or tracer for quantitative NMR, GC-MS, and LC-MS.
  • Molecular weight 172.27 and CAS 24493-21-8 for unambiguous identification.
  • High purity (98.0%) to support accurate quantitative analyses.
  • White crystalline powder; store sealed at 4 °C and protect from moisture.