Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(trimethylsilyl)propanoic acid-d4 (sodium) | 24493-21-8 | MFCD00002762 | 98.0% | 172.27 | C6H9D4NaO2Si | 5 MG
SDP

Catalog No. 5000284668
Encompass_Preferred

3-(Trimethylsilyl)propanoic acid-d4 sodium is a deuterium-labeled sodium salt of trimethylsilylpropionic acid supplied as a solid analytical standard for NMR and mass-spectrometry applications. It offers high isotopic labeling and chemical purity for reliable quantitative analyses.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity: 98.0%.
  • Molecular weight 172.27 and formula C6H9D4NaO2Si for accurate calculations.
  • Available in small pack sizes suitable for analytical standards.
  • Storage: sealed, away from moisture; 4°C recommended; in solvent: -80°C (6 months) or -20°C (1 month).
  • White to off-white solid form for easy handling and weighing.
  • For research use only; not for use in humans or patients.

Catalog No. 50-002-84668 Supplier Medchemexpress LLC Supplier No. HYW7026375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(Trimethylsilyl)propanoic acid-d4 sodium is a deuterium-labeled sodium salt of trimethylsilylpropionic acid supplied as a solid analytical standard for NMR and mass-spectrometry applications. It offers high isotopic labeling and chemical purity for reliable quantitative analyses.

  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • High chemical purity: 98.0%.
  • Molecular weight 172.27 and formula C6H9D4NaO2Si for accurate calculations.
  • Available in small pack sizes suitable for analytical standards.
  • Storage: sealed, away from moisture; 4°C recommended; in solvent: -80°C (6 months) or -20°C (1 month).
  • White to off-white solid form for easy handling and weighing.
  • For research use only; not for use in humans or patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.