Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Trimethylsilyl)propargyl alcohol

Catalog No. 3185585G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-855-85G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-855-85G Supplier Sigma Aldrich Supplier No. 3185585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Trimethylsilyl)propargyl alcohol

Specifications

CAS 5272-36-6
Density 0.865 g/mL (at 25°C (literature))
Boiling Point 76°C (11 mmHg)
Molecular Formula C6H12OSi
Linear Formula (CH3)3 SiC///CCH2OH
Refractive Index n20/D 1.451 (literature)
Quantity 250 mg
MDL Number MFCD00002913
Synonym 3-(Trimethylsilyl)-2-propyn-1-ol
Molecular Weight (g/mol) 128.24
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.