Learn More
3-(Trimethylsilyl)propionic-2,2,3,3-d4 acid sodium salt 98 atom% D, ≥98.0% (NMR), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 24493-21-8 |
| pH | 7.0 to 9.0 (1% in solution) |
| Packaging | Glass Bottle |
| Linear Formula | (CH3)3 SiCD2 CD2 CO2 Na |
| Quantity | 1 g |
| Formula Weight | 172.27 |
| Percent Purity | ≥98.0% (NMR) |
| Physical Form | Solid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.