Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trimethylsilyl)propionic acid-d4 sodium salt, 98 atom% D

Catalog No. AC432120010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432120010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432120010 Supplier Thermo Scientific Chemicals Supplier No. 432120010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24493-21-8 | C6H9D4NaO2Si | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24493-21-8
Molecular Formula C6H9D4NaO2Si
Molecular Weight (g/mol) 172.27
InChI Key OIIWPAYIXDCDNL-HGFPCDIYSA-M
Synonym sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate
PubChem CID 23688921
IUPAC Name sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate
SMILES C[Si](C)(C)CCC(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98 atom% D
Packaging Glass bottle
Linear Formula (CH3)3SiCD2CD2CO2Na
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 172.27
Percent Purity 98 atom% D
Physical Form Powder
Chemical Name or Material 3-(Trimethylsilyl)propionic acid-d4 sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 246-286-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.