Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Trimethylsilyl)propionic Acid, Sodium Salt 99+%, Thermo Scientific™

Catalog No. AC177390050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC177390050 5 g
AC177390010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177390050 Supplier Thermo Scientific Chemicals Supplier No. 177390050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37013-20-0
Molecular Formula C6H13NaO2Si
Molecular Weight (g/mol) 168.243
InChI Key OIIWPAYIXDCDNL-UHFFFAOYSA-M
Synonym sodium 3-trimethylsilyl propionate, sodium 3-trimethylsilyl propanoate, 3-trimethylsilyl propionic acid sodium salt, 3-trimethylsilyl-propionic acid sodium salt, sodium trimethylsilylpropanoate, sodium trimethylsilylpropionate, sodium 3-trimethylsilylpropionate, 3-trimethylsilyl propanoic acid sodium salt, 3-trimethylsilyl-propinoic acid sodium salt
PubChem CID 23666333
ChEBI CHEBI:85363
IUPAC Name sodium;3-trimethylsilylpropanoate
SMILES C[Si](C)(C)CCC(=O)[O-].[Na+]

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Linear Formula (CH3)3SiCH2CH2CO2Na
Quantity 5 g
Formula Weight 168.25
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material 3-(Trimethylsilyl)propionic acid, sodium salt, 99+%
EINECSNumber 253-308-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.