Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(Triphenylphosphonio)Propane-1-Sulfonate

Catalog No. 531665GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-166-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 53-166-5GF Supplier Sigma Aldrich Supplier No. 531665GF
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-(Triphenylphosphonio)propane-1-sulfonate is a zwitterionic, sulfonic acid-functionalized ionic liquid.3-(Triphenylphosphonio)propane-1-sulfonate has been used as a zwitterion to study its effect on the work function of indium tin oxide (ITO) in inverted polymer solar cells (PSCs). It can be used in the preparation of triphenyl-(3-sulfopropyl)phosphonium phosphotungstate, an efficient green catalyst for esterification reactions.

Specifications

CAS 7782-78-7
Melting Point 124°C to 128°C
Molecular Formula C23H21NO2
Linear Formula C23H21NO2
MDL Number MFCD01863736
Quantity 5 g
Molecular Weight (g/mol) 343.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.