Learn More
Medchemexpress LLC (3-{[Tris(hydroxymethyl)methyl]amino}propane-1-sulfonic acid) | 29915-38-6 | 98.0% | 50 G
SDP
Catalog No. 5000409966
Encompass_Preferred
TAPS is an aminosulfonate-based lysozyme stabilizer and connexin channel inhibitor. It helps maintain the native structure of lysozyme in aqueous solution at pH 7.0 and protects it from heat-induced denaturation. TAPS is also considered a critical compound for investigating the structure and function of connexin channels.
  • Maintains native structure of lysozyme in aqueous solution at pH 7.0
  • Protects lysozyme from heat-induced denaturation
  • Directly inhibits heteromeric connexin 32/connexin 26 channels (Cx32/Cx26)
  • Reduces connexin channel-mediated solute exchange in a recombinant liposome system
  • Critical compound for investigating connexin channel structure and function
Catalog No. 50-004-09966 Supplier Medchemexpress LLC Supplier No. HYD087750G
TAPS is an aminosulfonate-based lysozyme stabilizer and connexin channel inhibitor. It helps maintain the native structure of lysozyme in aqueous solution at pH 7.0 and protects it from heat-induced denaturation. TAPS is also considered a critical compound for investigating the structure and function of connexin channels.
  • Maintains native structure of lysozyme in aqueous solution at pH 7.0
  • Protects lysozyme from heat-induced denaturation
  • Directly inhibits heteromeric connexin 32/connexin 26 channels (Cx32/Cx26)
  • Reduces connexin channel-mediated solute exchange in a recombinant liposome system
  • Critical compound for investigating connexin channel structure and function