Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-[Tris(trimethylsiloxy)silyl]propyl methacrylate

Catalog No. 44613025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-613-025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-613-025ML Supplier Sigma Aldrich Supplier No. 44613025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-[Tris(trimethylsiloxy)silyl]propyl methacrylate

Specifications

CAS 17096-07-0
Density 0.918 g/mL (at 25°C)
Boiling Point 112°C to 115°C/0. (2 mmHg)
Molecular Formula C16H38O5Si4
Refractive Index n20/D 1.419 (literature)
Linear Formula H2C=C(CH3)CO2(CH2)3 Si[OSi(CH3)3]3
Quantity 25 mL
MDL Number MFCD00053871
Synonym Tris; (3-Methacryloyloxypropyl)tris(trimethylsiloxy)silane
Molecular Weight (g/mol) 422.81
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.