Learn More
Medchemexpress LLC 3-(Tritylthio)propylamine | 426828-04-8 | 95.0% | 333.49 | 50 MG
SDP
Catalog No. 5000401617
Encompass_Preferred
3-(Tritylthio)propylamine is a methyl-containing analogue of S-Trityl-L-cysteine, commonly found as a white to light yellow solid. It is classified as a drug derivative and has been referenced in studies concerning the structure-activity relationship of S-trityl-L-cysteine analogues as inhibitors of the human mitotic kinesin Eg5.
  • Methyl-containing analogue of S-Trityl-L-cysteine
  • Targeted as a drug derivative
  • Appears as a white to light yellow solid
  • Optimal storage conditions: 4°C, protected from light, under nitrogen. In solvent: -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-01617 Supplier Medchemexpress LLC Supplier No. HY40285350MG
3-(Tritylthio)propylamine is a methyl-containing analogue of S-Trityl-L-cysteine, commonly found as a white to light yellow solid. It is classified as a drug derivative and has been referenced in studies concerning the structure-activity relationship of S-trityl-L-cysteine analogues as inhibitors of the human mitotic kinesin Eg5.
  • Methyl-containing analogue of S-Trityl-L-cysteine
  • Targeted as a drug derivative
  • Appears as a white to light yellow solid
  • Optimal storage conditions: 4°C, protected from light, under nitrogen. In solvent: -80°C for 6 months or -20°C for 1 month.