Learn More
Selleck Chemical LLC 3-TYP 5mg 120241-79-4 3-(1H-1,2,3-triazol-4-yl) pyridine
SDP
Supplier:  Selleck Chemical LLC S86285MG
Encompass_Preferred
3-TYP (3-(1H-1,2,3-triazol-4-yl) pyridine) is a selective SIRT3 inhibitor which is selective for Sirt3 over Sirt1 and Sirt2. The IC50 values for SITR1, SIRT2, SIRT3 are 88 nM, 92 nM, 16 nM respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8035